missing translation for 'onlineSavingsMsg'
Learn More

Nonafluoro-tert-butyl Alcohol 98.0+%, TCI America™

Catalog No. N06925G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06925G 5 g
N06921G 1 g
N069225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N06925G Supplier TCI America Supplier No. N06925G

Chemical Identifiers

CAS 2378-02-1
Molecular Formula C4HF9O
Molecular Weight (g/mol) 236.04
MDL Number MFCD00042092
InChI Key XZNOAVNRSFURIR-UHFFFAOYSA-N
Synonym perfluoro-tert-butanol, nonafluoro-tert-butanol, perfluoro-tert-butyl alcohol, perfluoro-t-butanol, nonafluoro-tert-butyl alcohol, 2-propanol, 1,1,1,3,3,3-hexafluoro-2-trifluoromethyl, nonafluor-terc.butanol, 1,1,1,3,3,3-hexafluoro-2-trifluoromethyl propan-2-ol, 1,1,1,3,3,3-hexafluoro-2-trifluoromethyl-2-propanol, nonafluor-terc.butanol czech
PubChem CID 16924
IUPAC Name 1,1,1,3,3,3-hexafluoro-2-(trifluoromethyl)propan-2-ol
SMILES C(C(F)(F)F)(C(F)(F)F)(C(F)(F)F)O

Specifications

Color Colorless
Boiling Point 45°C
Quantity 5 g
Formula Weight 236.04
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Nonafluoro-tert-butyl Alcohol
RTECSNumber UB6452700
TSCA No