missing translation for 'onlineSavingsMsg'
Learn More

Nonafluoro-1-butanesulfonic Acid 98.0+%, TCI America™

Catalog No. N07095G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07095G 5 g
N070925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N07095G Supplier TCI America Supplier No. N07095G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 375-73-5
Molecular Formula C4HF9O3S
Molecular Weight (g/mol) 300.095
MDL Number MFCD01320794
InChI Key JGTNAGYHADQMCM-UHFFFAOYSA-N
Synonym Perfluoro-1-butanesulfonic Acid
PubChem CID 67815
IUPAC Name 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonic acid
SMILES C(C(C(F)(F)S(=O)(=O)O)(F)F)(C(F)(F)F)(F)F

Specifications

Color Yellow
Boiling Point 212°C
Quantity 5 g
UN Number 2586
Formula Weight 300.09
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Nonafluoro-1-butanesulfonic Acid
RTECSNumber EK5930000
TSCA Yes