missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nonaethylene Glycol Monomethyl Ether 93.0+%, TCI America™
$223.68 - $447.38
Chemical Identifiers
| CAS | 6048-68-6 |
|---|---|
| Molecular Formula | C19H40O10 |
| Molecular Weight (g/mol) | 428.52 |
| MDL Number | MFCD06797148 |
| InChI Key | VVHAVLIDQNWEKF-UHFFFAOYSA-N |
| Synonym | mPEG9-Alcohol |
| PubChem CID | 11339376 |
| ChEBI | CHEBI:59168 |
| IUPAC Name | 2,5,8,11,14,17,20,23,26-nonaoxaoctacosan-28-ol |
| SMILES | COCCOCCOCCOCCOCCOCCOCCOCCOCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
N0699500MG
|
TCI America
N0699500MG |
500 mg |
Each for $223.68
|
|
|||||
|
N06991G
|
TCI America
N06991G |
1 g |
Each for $447.38
|
|
|||||
Chemical Identifiers
| 6048-68-6 | |
| 428.52 | |
| VVHAVLIDQNWEKF-UHFFFAOYSA-N | |
| 11339376 | |
| 2,5,8,11,14,17,20,23,26-nonaoxaoctacosan-28-ol |
| C19H40O10 | |
| MFCD06797148 | |
| mPEG9-Alcohol | |
| CHEBI:59168 | |
| COCCOCCOCCOCCOCCOCCOCCOCCOCCO |
Specifications
| 6048-68-6 | |
| Yellow | |
| MFCD06797148 | |
| mPEG9-Alcohol | |
| COCCOCCOCCOCCOCCOCCOCCOCCOCCO | |
| 428.52 | |
| CHEBI:59168 | |
| ≥93.0% (GC) | |
| Nonaethylene Glycol Monomethyl Ether |
| 14°C | |
| C19H40O10 | |
| 500 mg | |
| VVHAVLIDQNWEKF-UHFFFAOYSA-N | |
| 2,5,8,11,14,17,20,23,26-nonaoxaoctacosan-28-ol | |
| 11339376 | |
| 428.52 | |
| Liquid |
Safety and Handling
EINECSNumber : (7)-0097
TSCA : No
Recommended Storage : Refrigerator