missing translation for 'onlineSavingsMsg'
Learn More

Nonadecylbenzene 98.0+%, TCI America™

Catalog No. p-7133762
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N03495G 5 g
N034925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N03495G Supplier TCI America Supplier No. N03495G
Only null left

Chemical Identifiers

CAS 29136-19-4
Molecular Formula C25H44
Molecular Weight (g/mol) 344.627
MDL Number MFCD00026692
InChI Key SHWJJBRTHGGZBE-UHFFFAOYSA-N
Synonym 1-Phenylnonadecane
PubChem CID 94400
IUPAC Name nonadecylbenzene
SMILES CCCCCCCCCCCCCCCCCCCC1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 344.63
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Nonadecylbenzene
EINECSNumber (3)-0021
TSCA Yes