missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nonadecanoic Acid 98.0+%, TCI America™
$329.94 - $572.27
Chemical Identifiers
| CAS | 646-30-0 |
|---|---|
| Molecular Formula | C19H38O2 |
| Molecular Weight (g/mol) | 298.511 |
| MDL Number | MFCD00002754 |
| InChI Key | ISYWECDDZWTKFF-UHFFFAOYSA-N |
| Synonym | n-nonadecanoic acid, nonadecylic acid, unii-h6m3vyc62p, h6m3vyc62p, fatty acids, c16-22, n-nonadecanoate, n-nonadecylate, nonadecyclic acid, c16-c22 alkylcarboxylic acid |
| PubChem CID | 12591 |
| ChEBI | CHEBI:39246 |
| IUPAC Name | nonadecanoic acid |
| SMILES | CCCCCCCCCCCCCCCCCCC(=O)O |
Chemical Identifiers
| 646-30-0 | |
| 298.511 | |
| ISYWECDDZWTKFF-UHFFFAOYSA-N | |
| 12591 | |
| nonadecanoic acid |
| C19H38O2 | |
| MFCD00002754 | |
| n-nonadecanoic acid, nonadecylic acid, unii-h6m3vyc62p, h6m3vyc62p, fatty acids, c16-22, n-nonadecanoate, n-nonadecylate, nonadecyclic acid, c16-c22 alkylcarboxylic acid | |
| CHEBI:39246 | |
| CCCCCCCCCCCCCCCCCCC(=O)O |
Specifications
| 646-30-0 | |
| White | |
| C19H38O2 | |
| 10 g | |
| ISYWECDDZWTKFF-UHFFFAOYSA-N | |
| nonadecanoic acid | |
| 12591 | |
| 298.51 | |
| Crystalline Powder |
| 68°C | |
| 238°C | |
| MFCD00002754 | |
| n-nonadecanoic acid, nonadecylic acid, unii-h6m3vyc62p, h6m3vyc62p, fatty acids, c16-22, n-nonadecanoate, n-nonadecylate, nonadecyclic acid, c16-c22 alkylcarboxylic acid | |
| CCCCCCCCCCCCCCCCCCC(=O)O | |
| 298.511 | |
| CHEBI:39246 | |
| ≥98.0% (GC) | |
| Nonadecanoic Acid |
Safety and Handling
EINECSNumber : (2)-0608
TSCA : Yes