missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nonadecanoic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 68°C |
| Color | White |
| Boiling Point | 238°C |
| Quantity | 10 g |
| Formula Weight | 298.51 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Nonadecanoic Acid |
Safety and Handling
| EINECSNumber | (2)-0608 |
| TSCA | Yes |