missing translation for 'onlineSavingsMsg'
Learn More

Nonadecane 99.0+%, TCI America™

Catalog No. N0282400G
Change view
Click to view available options
Quantity:
25 g
400 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0282400G 400 g
N028225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0282400G Supplier TCI America Supplier No. N0282400G

Chemical Identifiers

CAS 629-92-5
Molecular Formula C19H40
Molecular Weight (g/mol) 268.529
MDL Number MFCD00009012
InChI Key LQERIDTXQFOHKA-UHFFFAOYSA-N
Synonym n-nonadecane, nonadekan, unii-nmy21d3y5t, nmy21d3y5t, unknown branched fragment of phospholipid, ch3-ch2 17-ch3, unknown phospholipid fragment, nonadecane, analytical standard, upl
PubChem CID 12401
ChEBI CHEBI:32927
IUPAC Name nonadecane
SMILES CCCCCCCCCCCCCCCCCCC

Specifications

Color White-Yellow
Boiling Point 330°C
Quantity 400 g
Formula Weight 268.53
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Nonadecane
EINECSNumber (2)-0010
TSCA Yes