missing translation for 'onlineSavingsMsg'
Learn More

Nonadecane 99.5+%, TCI America™

Catalog No. S02915G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02915G Supplier TCI America Supplier No. S02915G

Chemical Identifiers

CAS 629-92-5
Molecular Formula C19H40
Molecular Weight (g/mol) 268.529
MDL Number MFCD00009012
InChI Key LQERIDTXQFOHKA-UHFFFAOYSA-N
Synonym n-nonadecane, nonadekan, unii-nmy21d3y5t, nmy21d3y5t, unknown branched fragment of phospholipid, ch3-ch2 17-ch3, unknown phospholipid fragment, nonadecane, analytical standard, upl
PubChem CID 12401
ChEBI CHEBI:32927
IUPAC Name nonadecane
SMILES CCCCCCCCCCCCCCCCCCC

Specifications

Color White
Quantity 5 g
Formula Weight 268.53
Percent Purity ≥99.5% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Nonadecane [Standard Material for GC]
EINECSNumber (2)-0010
TSCA Yes