missing translation for 'onlineSavingsMsg'
Learn More

Nonadecane, 99%

CAS: 629-92-5 | C19H40 | 268.51 g/mol

$87.25 - $215.05

Chemical Identifiers

CAS 629-92-5
Molecular Formula C19H40
Molecular Weight (g/mol) 268.51
MDL Number MFCD00009012
InChI Key LQERIDTXQFOHKA-UHFFFAOYSA-N
Synonym n-nonadecane, nonadekan, unii-nmy21d3y5t, nmy21d3y5t, unknown branched fragment of phospholipid, ch3-ch2 17-ch3, unknown phospholipid fragment, nonadecane, analytical standard, upl
PubChem CID 12401
ChEBI CHEBI:32927
IUPAC Name nonadecane
SMILES CCCCCCCCCCCCCCCCCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC129100250
SDS View Documents
Thermo Scientific Chemicals
129100250
25 g Glass bottle
Each for $87.25
Only null left
 
AC129101000
SDS View Documents
Thermo Scientific Chemicals
129101000
100 g Glass bottle
Each for $215.05
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

629-92-5
268.51
LQERIDTXQFOHKA-UHFFFAOYSA-N
12401
nonadecane
C19H40
MFCD00009012
n-nonadecane, nonadekan, unii-nmy21d3y5t, nmy21d3y5t, unknown branched fragment of phospholipid, ch3-ch2 17-ch3, unknown phospholipid fragment, nonadecane, analytical standard, upl
CHEBI:32927
CCCCCCCCCCCCCCCCCCC

Specifications

629-92-5
100.0
White
168°C
99%
C19H40
MFCD00009012
n-nonadecane, nonadekan, unii-nmy21d3y5t, nmy21d3y5t, unknown branched fragment of phospholipid, ch3-ch2 17-ch3, unknown phospholipid fragment, nonadecane, analytical standard, upl
LQERIDTXQFOHKA-UHFFFAOYSA-N
nonadecane
12401
CHEBI:32927
99%
Nonadecane
98.5
32.0°C to 34.0°C
330.0°C
Authentic
Glass bottle
CH3(CH2)17CH3
25 g
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
CCCCCCCCCCCCCCCCCCC
268.51
3.6 mPa.s (40°C)
268.51
Crystalline Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.

GHS Signal Word: Danger

EINECSNumber : 211-116-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only