missing translation for 'onlineSavingsMsg'
Learn More

Contec Non-Sterile USP Grade Isopropanol

Catalog No. 19999857
Change view
Click to view available options
Sterility:
Non-sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
19999857 Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19999857 Supplier Contec Supplier No. SB128100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Non-sterile USP grade isopropanol for wiping gloved hands during gowning and wiping products before pass-through.

Contec Isopropanol contains USP grade isopropanol (IPA). Filtered to 0.2μm, bottles are double bagged to assure product quality.

  • Each bottle has a lot number code and expiration date for easy record keeping.
  • Each bottle is induction sealed and capped to eliminate the occurrence of leakage during shipping.
  • Product applications include surface cleaning, wiping gloved hands in sterile suites or during gowning, and wiping down products and equipment for pass through to controlled environments.

Specifications

Product Type Isopropyl Alcohol
Form Clear Liquid
pH 7
Scent Unscented
Sterility Non-sterile
Appearance Clear
Autoclavable Not Autoclavable
Color Clear
Composition 99% IPA
Disposable Single-use
Filtered Filtered
Flammable Yes
Show More Show Less