missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Henke-Sass, Wolf Non-Sterile 2-Part Syringes
These syringes feature a 2-part construction containing no rubber and eliminating the need for silicone oil. Made from polypropylene and polyethylene.
Supplier: Henke-Sass, Wolf AB1
Description
- Disposable Syringes
- 2-Part Syringe
- Unsterile
- Polypropylene Barrel
- Polyethylene Plunger
- Single-use
- Latex Free
- ISO
- Chromatography
- Laboratory Use
Specifications
| Not Autoclavable | |
| Polyethylene (Plunger), Polypropylene (Barrel) | |
| 7000 Syringes | |
| Single-use | |
| 1 mL | |
| Yes |
| ISO 13485 | |
| Clear | |
| Yes | |
| Bulk | |
| Leur Slip | |
| Non-sterile |