Learn More
Fisherbrand™ Round-Bottom Polypropylene Test Tubes Without Cap
Fisherbrand™'s round bottom tubes provide reliable containment for various laboratory fluid samples and preparations.
 View more versions of this product
Catalog No. FB149569A
Only null left
Description
Description
  • Ideal for use in laboratory protocols requiring round bottom tubes
  • Ensures reliable handling of liquid samples
  • Polypropylene tubes are best suited for applications requiring greater thermal and chemical stability, while polystyrene tubes offer high optical clarity
  • Available with and without a snap cap - dual-position snap caps offer both vented and fully closed positions.
  • 3,000 RCF rating for polypropylene tubes, 1,400 RCF rating for polystyrene tubes
  • Dimensions and volumes are approximate
Specifications
Chemical Identifiers
Translucent
Single-use
Not Included
Polypropylene
Fisherbrand
Test Tube
Specifications
Translucent
100 mm
Single-use
Sterile
Yes
Test Tube
14 mL
Polypropylene
Blister Pack
Fisherbrand
Not Included
Product Suggestions
Product Suggestions