Learn More
Globe Scientific Non-Graduated Screwtop Storage Tubes
Description
- For the storage and transportation of biological material
- Manufactured from non-toxic polypropylene (PP) to offer strength and clarity
- External threads provide a smooth and uniform inner surface which reduces the risk of contamination
- Screw caps are also constructed of polypropylene to provide the same coefficient of expansion as the tubes
- Screw caps sold separately (21-200-103)
- Round bottom
Order Info
Screwcaps (21200103) sold separately
Specifications
Specifications
| Color | Natural |
| Capacity (Metric) | 5 mL |
| Bottom Shape | Round |
| Material | Polypropylene |
| Thread Style | External |
| Quantity | 1000/Bag |
| Closure Type | Screw Cap (Not Included) |
| Product Type | Storage Tube |