missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nomega-Nitro-L-arginine Methyl Ester Hydrochloride 98.0+%, TCI America™
$197.46 - $525.92
Chemical Identifiers
| CAS | 51298-62-5 |
|---|---|
| Molecular Formula | C7H16ClN5O4 |
| Molecular Weight (g/mol) | 269.69 |
| MDL Number | MFCD00039052,MFCD00133613 |
| InChI Key | QBNXAGZYLSRPJK-JEDNCBNOSA-N |
| Synonym | h-arg no2-ome.hcl, lname hydrochloride, l-name hydrochloride, ng-nitro-l-arginine methyl ester hydrochloride, nomega-nitro-l-arginine methyl ester hydrochloride, s-methyl 2-amino-5-3-nitroguanidino pentanoate hydrochloride, l-name, hcl, ng-no2-l-arg-ome, nomega-no2-l-arg-ome |
| PubChem CID | 135193 |
| IUPAC Name | hydrogen methyl (2S)-2-amino-5-{[amino(nitroamino)methylidene]amino}pentanoate chloride |
| SMILES | [H+].[Cl-].COC(=O)[C@@H](N)CCCN=C(N)N[N+]([O-])=O |
Chemical Identifiers
| 51298-62-5 | |
| 269.69 | |
| QBNXAGZYLSRPJK-JEDNCBNOSA-N | |
| 135193 | |
| [H+].[Cl-].COC(=O)[C@@H](N)CCCN=C(N)N[N+]([O-])=O |
| C7H16ClN5O4 | |
| MFCD00039052,MFCD00133613 | |
| h-arg no2-ome.hcl, lname hydrochloride, l-name hydrochloride, ng-nitro-l-arginine methyl ester hydrochloride, nomega-nitro-l-arginine methyl ester hydrochloride, s-methyl 2-amino-5-3-nitroguanidino pentanoate hydrochloride, l-name, hcl, ng-no2-l-arg-ome, nomega-no2-l-arg-ome | |
| hydrogen methyl (2S)-2-amino-5-{[amino(nitroamino)methylidene]amino}pentanoate chloride |
Specifications
| 159°C | |
| White | |
| MFCD00039052,MFCD00133613 | |
| QBNXAGZYLSRPJK-JEDNCBNOSA-N | |
| hydrogen methyl (2S)-2-amino-5-{[amino(nitroamino)methylidene]amino}pentanoate chloride | |
| 269.69 | |
| 269.69 | |
| Crystalline Powder |
| 51298-62-5 | |
| C7H16ClN5O4 | |
| h-arg no2-ome.hcl, lname hydrochloride, l-name hydrochloride, ng-nitro-l-arginine methyl ester hydrochloride, nomega-nitro-l-arginine methyl ester hydrochloride, s-methyl 2-amino-5-3-nitroguanidino pentanoate hydrochloride, l-name, hcl, ng-no2-l-arg-ome, nomega-no2-l-arg-ome | |
| [H+].[Cl-].COC(=O)[C@@H](N)CCCN=C(N)N[N+]([O-])=O | |
| 5 g | |
| 135193 | |
| ≥98.0% (HPLC,T) | |
| Nomega-Nitro-L-arginine Methyl Ester Hydrochloride |
Safety and Handling
TSCA : No