missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nomega-Nitro-L-arginine Methyl Ester Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 159°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 269.69 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Grade | Research |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Nomega-Nitro-L-arginine Methyl Ester Hydrochloride |
Safety and Handling
| TSCA | No |