missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nobiletin 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 138°C |
| Color | White-Yellow |
| Quantity | 100 mg |
| Synonym | 3′,4′,5,6,7,8-Hexamethoxyflavone |
| Formula Weight | 402.40 |
| Percent Purity | ≥95.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Nobiletin |
Safety and Handling
| RTECSNumber | DJ3052200 |
| TSCA | No |
| Recommended Storage | Freezer |