missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals NOB, 98%

Catalog No. AAJ67360MF
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67360MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67360MF Supplier Thermo Scientific Chemicals Supplier No. J67360MF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 55894-52-5 | C15H19NO6, C15H19NO6 | 309.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55894-52-5
Molecular Formula C15H19NO6
Synonym 4-Nitro-3-(octanoyloxy)benzoic acid;

Specifications

Quantity 50 mg
Description Off-white
Formula Weight 309.32
Product Type NOB, 98%
Form Solid
Assay 98%

RUO – Research Use Only