Learn More
Brady™ No-Handle Valve Lockout with Cable for Ball and Gate Valves, Clear
Catalog No. 17985485
Change view
Click to view available options
Material:
Glass-filled Nylon, PVC
Catalog No. Material
17985485 Glass-filled Nylon, PVC
1 options
Catalog No. 17985485 Supplier Brady™ Supplier No. 178769
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
No handle, no problem - experience the convenience of cable lockouts for handle-off ball, gate, and pneumatic valves with this versatile lockout.
  • Eliminate the hassle of managing multiple devices with this innovative design, optimized to fit most valve types and sizes.
  • Easy to apply - simply place the cap, thread the cable, close the door, pull tight, and secure with a padlock.
  • Stays securely in place - durable cam wheels hold the cable tightly, and a closable protective cam door prevents loosening or tampering.
  • The cable can be tightened but never loosened once the cam door is closed, ensuring reliable lockout protection.
  • Two interchangeable rubber bases provide a snug fit to further prevent tampering and ensure secure attachment.
  • The 3 ft. sheathed metal cable is built to withstand heavy use and tough conditions.
  • Four lock holes, including a hasp-compatible bottom hole, provide secure locking points for multiple workers.
  • The clear cover allows quick visual verification of the valve's on/off position, enhancing safety checks.
Specifications
Product Type Valve Lockout with Cable
Color Red
Material Glass-filled Nylon, PVC