missing translation for 'onlineSavingsMsg'
Learn More

[Nle4, D-Phe7] alpha-Melanocyte Stimulating Hormone amide trifluoroacetate salt, Thermo Scientific Chemicals

Catalog No. AAJ66191MCR
Change view
Click to view available options
Quantity:
1 mg
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ66191MCR 1 mg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66191MCR Supplier Thermo Scientific Chemicals Supplier No. J66191MCR
Only null left

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75921-69-6
Molecular Formula C78H111N21O19
MDL Number MFCD00151715
Synonym N-Acetyl-Ser-Tyr-Ser-Nle-Glu-His-DPhe-Arg-Trp-Gly-Lys-Pro-Val-NH{2},Melanotan

Specifications

Packaging Plastic bottle
Quantity 1 mg
Formula Weight 1647
Physical Form Solid
Chemical Name or Material alpha-Melanocyte Stimulating Hormone amide trifluoroacetate salt
RTECSNumber OS1830200
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only