missing translation for 'onlineSavingsMsg'
Learn More

Nitrosonium tetrafluoroborate, 97%

Catalog No. p-3734681
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC174390050 5 g Plastic bottle
AC174390250 25 g Plastic bottle
AC174391000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC174390050 Supplier Thermo Scientific Chemicals Supplier No. 174390050

CAS: 14635-75-7 | BF4HNO2 | 133.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent

Chemical Identifiers

CAS 14635-75-7
Molecular Formula BF4HNO2
Molecular Weight (g/mol) 133.82
MDL Number MFCD00011433
InChI Key XYRQURYIJKJVSF-UHFFFAOYSA-N
Synonym nitrosonium tetrafluoroborate, nitrosyl tetrafluoroborate, nitrosyltetrafluoroborate, nobf4, nitrilooxonium tetrafluoroborate, azanylidyneoxidanium tetrafluoroborate
PubChem CID 11137142
IUPAC Name azanylidyneoxidanium;tetrafluoroborate
SMILES [O-][NH+]=O.F[B-](F)(F)F

Specifications

Melting Point 250.0°C
Color White
CAS Min % 96.0
CAS Max % 100.0
Physical Form Crystalline Powder and/or Crystals
Quantity 5 g
Assay Percent Range 96% min. (ex F)
Linear Formula NOBF4
Merck Index 15, 6736
Formula Weight 116.81
Percent Purity 97%
Packaging Plastic bottle
Specific Gravity 2.185
Density 2.1850g/mL
Chemical Name or Material Nitrosonium tetrafluoroborate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 238-679-2

RUO – Research Use Only