missing translation for 'onlineSavingsMsg'
Learn More

Nitrosonium hexafluorophosphate, 96%

Catalog No. p-7049181
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1314906 5 g
AA1314914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1314906 Supplier Thermo Scientific Chemicals Supplier No. 01314906
Only null left

CAS: 16921-91-8 | F6NOP | 174.97 g/mol

Nitrosonium hexafluorophosphate is used as initiator in the polymerization reactions of 1,2-epoxides such as styrene oxide, cyclohexene oxide and 1,2-epoxybutane. Further, it is involved in the polymerization of styrene, n-butyl vinyl ether and tetrahydrofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16921-91-8
Molecular Formula F6NOP
Molecular Weight (g/mol) 174.97
MDL Number MFCD00040324
InChI Key SAKPNYRUBHJGAR-UHFFFAOYSA-N
Synonym nitrosonium hexafluorophosphate, nitrilooxonium hexafluorophosphate, nopf6, nitrosyl hexafluorophosphate, nitrosonium hexafluorophosphat, nitrosylhexafluorophosphate, azanylidyneoxidanium hexafluorophosphate, hexafluoro-$l^ 5-phosphanuide; nitrosonium
PubChem CID 10910083
IUPAC Name azanylidyneoxidanium;hexafluorophosphate
SMILES N#[O+].F[P-](F)(F)(F)(F)F

Specifications

Physical Form Crystalline
Quantity 5 g
Linear Formula NOPF6
UN Number UN3260
Solubility Information Soluble in acetonitrile.
Formula Weight 174.96
Percent Purity 96%
Odor Odorless
Sensitivity Moisture sensitive
Chemical Name or Material Nitrosonium hexafluorophosphate
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 240-980-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only