missing translation for 'onlineSavingsMsg'
Learn More

Nitrosonium hexafluorophosphate, 95%

Catalog No. p-3523523
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC383730050 5 g Plastic bottle
AC383730250 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC383730050 Supplier Thermo Scientific Chemicals Supplier No. 383730050
Only null left

CAS: 16921-91-8 | F6NOP | 174.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16921-91-8
Molecular Formula F6NOP
Molecular Weight (g/mol) 174.97
MDL Number MFCD00040324
InChI Key SAKPNYRUBHJGAR-UHFFFAOYSA-N
Synonym nitrosonium hexafluorophosphate, nitrilooxonium hexafluorophosphate, nopf6, nitrosyl hexafluorophosphate, nitrosonium hexafluorophosphat, nitrosylhexafluorophosphate, azanylidyneoxidanium hexafluorophosphate, hexafluoro-$l^ 5-phosphanuide; nitrosonium
PubChem CID 10910083
IUPAC Name azanylidyneoxidanium;hexafluorophosphate
SMILES N#[O+].F[P-](F)(F)(F)(F)F

Specifications

Color Colorless or White
CAS Min % 94.0
CAS Max % 100.0
Physical Form Crystals
Quantity 5 g
Assay Percent Range 94% min
Solubility Information Solubility in water: decomposes
Formula Weight 174.97
Percent Purity 95%
Packaging Plastic bottle
Chemical Name or Material Nitrosonium hexafluorophosphate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 240-980-9

RUO – Research Use Only