missing translation for 'onlineSavingsMsg'
Learn More

Nitrosobenzene 98.0+%, TCI America™

$145.40 - $397.05

Chemical Identifiers

CAS 586-96-9
Molecular Formula C6H5NO
Molecular Weight (g/mol) 107.112
MDL Number MFCD00002059
InChI Key NLRKCXQQSUWLCH-UHFFFAOYSA-N
Synonym benzene, nitroso, nitroso-benzene, unii-zi9w9e8g2z, nob, zi9w9e8g2z, chembl98797, nitroso benzene, keto-aniline, nitrosobenzene, acmc-209m6j
PubChem CID 11473
ChEBI CHEBI:27986
IUPAC Name nitrosobenzene
SMILES C1=CC=C(C=C1)N=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N03675G
View Documents
TCI America
N03675G
5 g
Each for $145.40
Only null left
 
N036725G
View Documents
TCI America
N036725G
25 g
Each for $397.05
Only null left
 
Description

Description

Specifications

Chemical Identifiers

586-96-9
107.112
NLRKCXQQSUWLCH-UHFFFAOYSA-N
11473
nitrosobenzene
C6H5NO
MFCD00002059
benzene, nitroso, nitroso-benzene, unii-zi9w9e8g2z, nob, zi9w9e8g2z, chembl98797, nitroso benzene, keto-aniline, nitrosobenzene, acmc-209m6j
CHEBI:27986
C1=CC=C(C=C1)N=O

Specifications

586-96-9
White-Yellow
C6H5NO
5 g
benzene, nitroso, nitroso-benzene, unii-zi9w9e8g2z, nob, zi9w9e8g2z, chembl98797, nitroso benzene, keto-aniline, nitrosobenzene, acmc-209m6j
C1=CC=C(C=C1)N=O
107.112
CHEBI:27986
≥98.0% (GC)
Nitrosobenzene
66°C
57°C
MFCD00002059
2811
NLRKCXQQSUWLCH-UHFFFAOYSA-N
nitrosobenzene
11473
107.11
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2752

RTECSNumber : DA6497525

TSCA : Yes

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'