missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitronium tetrafluoroborate, 96%
Description
Nitronium tetrafluoroborate has been shown to be a versatile nitrating agent for nitrogen compounds, giving the corresponding N-nitro derivative when reacted with secondary aliphatic amines, an acyl aliphatic amine, a carbamate ester: a diacyl amine and primary amides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Nitronium tetrafluoroborate has been shown to be a versatile nitrating agent for nitrogen compounds, giving the corresponding N-nitro derivative when reacted with secondary aliphatic amines, an acyl aliphatic amine, a carbamate ester: a diacyl amine and primary amides.
Solubility
Decomposes in water.
Notes
Moisture sensitive. Store away from oxidizing agents, bases, water/moisture and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep it refrigerated.
Nitronium tetrafluoroborate has been shown to be a versatile nitrating agent for nitrogen compounds, giving the corresponding N-nitro derivative when reacted with secondary aliphatic amines, an acyl aliphatic amine, a carbamate ester: a diacyl amine and primary amides.
Solubility
Decomposes in water.
Notes
Moisture sensitive. Store away from oxidizing agents, bases, water/moisture and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep it refrigerated.
Specifications
Specifications
| Melting Point | ∽240°C (decomposition) |
| Quantity | 100 g |
| Sensitivity | Moisture sensitive |
| Linear Formula | NO2BF4 |
| UN Number | UN3260 |
| Solubility Information | Decomposes in water. |
| IUPAC Name | nitronium;tetrafluoroborate |
| Formula Weight | 132.81 |
| Percent Purity | 96% |
| Chemical Name or Material | Nitronium tetrafluoroborate |
Safety and Handling
| Hazard Category | H314-H334 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P280-P285-P301+P330+P331-P303+P361+P353-P304+P341-P305+P351+P338-P310-P342+P311-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S. |
| EINECSNumber | 237-533-5 |
| RTECSNumber | QX8620000 |
| TSCA | Yes |
| Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only