missing translation for 'onlineSavingsMsg'
Learn More

Nitronium tetrafluoroborate, 96%

Catalog No. AAB2016706
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2016706 5 g
AAB2016714 25 g
AAB2016722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2016706 Supplier Thermo Scientific Chemicals Supplier No. B2016706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13826-86-3 | BF4NO2 | 132.81 g/mol

Nitronium tetrafluoroborate has been shown to be a versatile nitrating agent for nitrogen compounds, giving the corresponding N-nitro derivative when reacted with secondary aliphatic amines, an acyl aliphatic amine, a carbamate ester: a diacyl amine and primary amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nitronium tetrafluoroborate has been shown to be a versatile nitrating agent for nitrogen compounds, giving the corresponding N-nitro derivative when reacted with secondary aliphatic amines, an acyl aliphatic amine, a carbamate ester: a diacyl amine and primary amides.

Solubility
Decomposes in water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, water/moisture and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep it refrigerated.

Chemical Identifiers

CAS 13826-86-3
Molecular Formula BF4NO2
Molecular Weight (g/mol) 132.81
MDL Number MFCD00011432
InChI Key XYRQURYIJKJVSF-UHFFFAOYSA-N
Synonym nitronium tetrafluoroborate, nitronium ion tetrafluoroborate, nitronium tetrafluoroboron, nitroniumtetrafluoroborate, nitronium.tetrafluoroborate, nitronium tetrafluoroborate, 0.3-0.5m solution in sulfolane
PubChem CID 11073463
SMILES [O-][NH+]=O.F[B-](F)(F)F

Specifications

Melting Point ∽240°C (decomposition)
Quantity 5 g
Linear Formula NO2BF4
UN Number UN3260
Solubility Information Decomposes in water.
IUPAC Name nitronium;tetrafluoroborate
Formula Weight 132.81
Percent Purity 96%
Sensitivity Moisture sensitive
Chemical Name or Material Nitronium tetrafluoroborate
Hazard Category H314-H334
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P285-P301+P330+P331-P303+P361+P353-P304+P341-P305+P351+P338-P310-P342+P311-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 237-533-5
RTECSNumber QX8620000
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only