missing translation for 'onlineSavingsMsg'
Learn More

Nitronium tetrafluoroborate, 90%

Catalog No. AA3637414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3637414 25 g
AA3637406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3637414 Supplier Thermo Scientific Chemicals Supplier No. 03637414

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13826-86-3 | BF4NO2 | 132.81 g/mol

Ingold's method of competitive nitration has been used in order to determine relative reactivities of alkylbenzenes and benzene in homogeneous mononitrations with nitronium tetrafluoroborate in tetramethylene sulfone solutions. Oxidation of both exo-and endo-2-bromonorbornane with nitronium tetrafluoroborate in acetonitrile afforded N-exo-2-norbornylacetamide. Used in the synthesis and characterization of new polyhydroxyorganocarboxylated fullerene derivatives through the electrophilic addition of nitronium tetrafluoroborate onto fullerenes in the presence of arenecarboxylic acid in a nonaqueous medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13826-86-3
Molecular Formula BF4NO2
Molecular Weight (g/mol) 132.81
MDL Number MFCD00011432
InChI Key XYRQURYIJKJVSF-UHFFFAOYSA-N
Synonym nitronium tetrafluoroborate, nitronium ion tetrafluoroborate, nitronium tetrafluoroboron, nitroniumtetrafluoroborate, nitronium.tetrafluoroborate, nitronium tetrafluoroborate, 0.3-0.5m solution in sulfolane
PubChem CID 11073463
IUPAC Name nitronium;tetrafluoroborate
SMILES [O-][NH+]=O.F[B-](F)(F)F

Specifications

Melting Point 240°C (dec.)
Color White
Physical Form Crystalline
Quantity 25 g
Sensitivity Moisture sensitive
Linear Formula NO2BF4
UN Number UN3260
Solubility Information Decomposes in water.
Formula Weight 132.81
Percent Purity 90%
Chemical Name or Material Nitronium tetrafluoroborate
Show More Show Less
Hazard Category H314-H334
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P285-P301+P330+P331-P303+P361+P353-P304+P341-P305+P351+P338-P310-P342+P311-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 237-533-5
RTECSNumber QX8620000
TSCA Yes
storageNote1 2°C to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only