Learn More
Nitronium tetrafluoroborate, 90%
CAS: 13826-86-3 | BF4NO2 | 132.81 g/mol
$118.27 - $331.88
Chemical Identifiers
| CAS | 13826-86-3 |
|---|---|
| Molecular Formula | BF4NO2 |
| Molecular Weight (g/mol) | 132.81 |
| MDL Number | MFCD00011432 |
| InChI Key | XYRQURYIJKJVSF-UHFFFAOYSA-N |
| Synonym | nitronium tetrafluoroborate, nitronium ion tetrafluoroborate, nitronium tetrafluoroboron, nitroniumtetrafluoroborate, nitronium.tetrafluoroborate, nitronium tetrafluoroborate, 0.3-0.5m solution in sulfolane |
| PubChem CID | 11073463 |
| IUPAC Name | nitronium;tetrafluoroborate |
| SMILES | [O-][NH+]=O.F[B-](F)(F)F |
Description
Ingold's method of competitive nitration has been used in order to determine relative reactivities of alkylbenzenes and benzene in homogeneous mononitrations with nitronium tetrafluoroborate in tetramethylene sulfone solutions. Oxidation of both exo-and endo-2-bromonorbornane with nitronium tetrafluoroborate in acetonitrile afforded N-exo-2-norbornylacetamide. Used in the synthesis and characterization of new polyhydroxyorganocarboxylated fullerene derivatives through the electrophilic addition of nitronium tetrafluoroborate onto fullerenes in the presence of arenecarboxylic acid in a nonaqueous medium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13826-86-3 | |
| 132.81 | |
| XYRQURYIJKJVSF-UHFFFAOYSA-N | |
| 11073463 | |
| [O-][NH+]=O.F[B-](F)(F)F |
| BF4NO2 | |
| MFCD00011432 | |
| nitronium tetrafluoroborate, nitronium ion tetrafluoroborate, nitronium tetrafluoroboron, nitroniumtetrafluoroborate, nitronium.tetrafluoroborate, nitronium tetrafluoroborate, 0.3-0.5m solution in sulfolane | |
| nitronium;tetrafluoroborate |
Specifications
| 240°C (dec.) | |
| White | |
| 5 g | |
| NO2BF4 | |
| UN3260 | |
| Decomposes in water. | |
| [O-][NH+]=O.F[B-](F)(F)F | |
| 132.81 | |
| 132.81 | |
| Moisture sensitive |
| 13826-86-3 | |
| Crystalline | |
| BF4NO2 | |
| MFCD00011432 | |
| nitronium tetrafluoroborate, nitronium ion tetrafluoroborate, nitronium tetrafluoroboron, nitroniumtetrafluoroborate, nitronium.tetrafluoroborate, nitronium tetrafluoroborate, 0.3-0.5m solution in sulfolane | |
| XYRQURYIJKJVSF-UHFFFAOYSA-N | |
| nitronium;tetrafluoroborate | |
| 11073463 | |
| 90% | |
| Nitronium tetrafluoroborate |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P285-P301+P330+P331-P303+P361+P353-P304+P341-P305+P351+P338-P310-P342+P311-P363-P501c
H314-H334
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 237-533-5
RTECSNumber : QX8620000
TSCA : Yes
storageNote1 : 2°C to 8°C
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only