missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitro Blue Tetrazolium 98.0+%, TCI America™
$40.44 - $216.92
Chemical Identifiers
| CAS | 298-83-9 |
|---|---|
| Molecular Formula | C40H30Cl2N10O6 |
| Molecular Weight (g/mol) | 817.644 |
| MDL Number | MFCD00012159 |
| InChI Key | FSVCQIDHPKZJSO-UHFFFAOYSA-L |
| Synonym | 2,2′C-Di-p-nitrophenyl-5,5′C-diphenyl-(3,3′C-dimethoxy)-4,4′C-bisphenyleneditetrazolium Chloride, NBT |
| PubChem CID | 9281 |
| ChEBI | CHEBI:9505 |
| IUPAC Name | 2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride |
| SMILES | COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D0844100MG
|
TCI America
D0844100MG |
100 mg |
Each for $40.44
|
|
|||||
|
D08441G
|
TCI America
D08441G |
1 g |
Each for $216.92
|
|
|||||
Chemical Identifiers
| 298-83-9 | |
| 817.644 | |
| FSVCQIDHPKZJSO-UHFFFAOYSA-L | |
| 9281 | |
| 2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride |
| C40H30Cl2N10O6 | |
| MFCD00012159 | |
| 2,2′C-Di-p-nitrophenyl-5,5′C-diphenyl-(3,3′C-dimethoxy)-4,4′C-bisphenyleneditetrazolium Chloride, NBT | |
| CHEBI:9505 | |
| COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-] |
Specifications
| 223°C | |
| C40H30Cl2N10O6 | |
| 2,2′C-Di-p-nitrophenyl-5,5′C-diphenyl-(3,3′C-dimethoxy)-4,4′C-bisphenyleneditetrazolium Chloride, NBT | |
| COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-] | |
| 817.644 | |
| CHEBI:9505 | |
| 100 mg | |
| Nitro Blue Tetrazolium [for Biochemical Research] |
| 298-83-9 | |
| MFCD00012159 | |
| FSVCQIDHPKZJSO-UHFFFAOYSA-L | |
| 2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride | |
| 9281 | |
| ≥98.0% (HPLC,T) | |
| Crystal/Powder |
Safety and Handling
RTECSNumber : XF8045000
TSCA : Yes
Recommended Storage : Freezer