missing translation for 'onlineSavingsMsg'
Learn More

Nitro Blue Tetrazolium 98.0+%, TCI America™

Catalog No. D0844100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0844100MG 100 mg
D08441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0844100MG Supplier TCI America Supplier No. D0844100MG

Chemical Identifiers

CAS 298-83-9
Molecular Formula C40H30Cl2N10O6
Molecular Weight (g/mol) 817.644
MDL Number MFCD00012159
InChI Key FSVCQIDHPKZJSO-UHFFFAOYSA-L
Synonym 2,2′C-Di-p-nitrophenyl-5,5′C-diphenyl-(3,3′C-dimethoxy)-4,4′C-bisphenyleneditetrazolium Chloride, NBT
PubChem CID 9281
ChEBI CHEBI:9505
IUPAC Name 2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride
SMILES COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-]

Specifications

Melting Point 223°C
Percent Purity ≥98.0% (HPLC,T)
Quantity 100 mg
Physical Form Crystal/Powder
Chemical Name or Material Nitro Blue Tetrazolium [for Biochemical Research]
RTECSNumber XF8045000
TSCA Yes
Recommended Storage Freezer