missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nitro Blue Tetrazolium Chloride monohydrate, 90%

Catalog No. AC128590010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC128590010 1 g
AC128590050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC128590010 Supplier Thermo Scientific Chemicals Supplier No. 128590010

CAS: 298-83-9 | C40H30Cl2N10O6 | 817.644 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Nitro Blue Tetrazolium Chloride monohydrate
CAS 298-83-9, 67-56-1, 7732-18-5
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Molecular Formula C40H30Cl2N10O6
MDL Number MFCD00150013
Synonym Nitro BT, Nitro blue tetrazolium
InChI Key FSVCQIDHPKZJSO-UHFFFAOYSA-L
SMILES COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-]
Infrared Spectrum Authentic
IUPAC Name 2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride
Molecular Weight (g/mol) 817.644
PubChem CID 9281
ChEBI CHEBI:9505
Formula Weight 867.69
Loss on Drying 10% max. (105°C)
Percent Purity ≥88%
Packaging Glass bottle
Color Yellow
Quantity 1 g
Physical Form Powder
Show More Show Less

RUO – Research Use Only