missing translation for 'onlineSavingsMsg'
Learn More

Ansell BioClean-D™ Disposable Long Sleeve Cover

Catalog No. 19668340
Change view
Click to view available options
Sterility:
Sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
19668340 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19668340 Supplier Ansell Supplier No. SBDSCL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Extra long length sleeve covers are constructed from antistatic, low-linting CleanTough™ material and are ISO Class 4 compatible.

  • Protective sleeves available in sterile or non-sterile
  • Low linting, anti-static CleanTough™ fabric
  • Elasticated arms and cuffs
  • ISO Class 4 and EU GMP Grade A compatible
  • Universal size
  • Overall length: 500 mm
  • Applications include cleanroom, pharma, controlled environments, 503B facilities/compounding, manufacturing, industrial

Specifications

Product Type Controlled Environment Sleeve
Size Standard
Material Double Laminate Microporous Fabric
Color White
Sterility Sterile
Disposable Single-use
Closure Type Elastic Ends
Model BDSC
For Use With (Application) Cleanroom, Clinical, Medical, Laboratory
Certifications/Compliance ASTM D6978-05, ASTM F739-12, ISO 9001:2015
Certifications/Compliance (Europe) (EU)2016/425, EN ISO 13034:2005+A1:2009, EN ISO 14325, EN 1149-5:2008, EN 530, EN 7854, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2
Length (English) 19.6 in.
Length (Metric) 500 mm
Cleanroom Class Class 10 (ISO 4)
Show More Show Less