missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrilotris(methylenephosphonic Acid) (ca. 50% in Water, ca. 2.2mol/L), TCI America™
$89.48 - $322.48
Chemical Identifiers
| CAS | 6419-19-8 |
|---|---|
| Molecular Formula | C3H12NO9P3 |
| Molecular Weight (g/mol) | 299.048 |
| MDL Number | MFCD00002138 |
| InChI Key | YDONNITUKPKTIG-UHFFFAOYSA-N |
| Synonym | nitrilotris methylene triphosphonic acid, tris phosphonomethyl amine, ferrofos 509, aminotrimethylene phosphonic acid, dowell l 37, dequest 2000, nitrilotrimethylphosphonic acid, nitrilotris methylenephosphonic acid, aminotris methylphosphonic acid, aminotri methylenephosphonic acid |
| PubChem CID | 16698 |
| IUPAC Name | [bis(phosphonomethyl)amino]methylphosphonic acid |
| SMILES | C(N(CP(=O)(O)O)CP(=O)(O)O)P(=O)(O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
N047425ML
|
TCI America
N047425ML |
25 mL |
Each for $89.48
|
|
|||||
|
N0474500ML
|
TCI America
N0474500ML |
500 mL |
Each for $322.48
|
|
|||||
Chemical Identifiers
| 6419-19-8 | |
| 299.048 | |
| YDONNITUKPKTIG-UHFFFAOYSA-N | |
| 16698 | |
| C(N(CP(=O)(O)O)CP(=O)(O)O)P(=O)(O)O |
| C3H12NO9P3 | |
| MFCD00002138 | |
| nitrilotris methylene triphosphonic acid, tris phosphonomethyl amine, ferrofos 509, aminotrimethylene phosphonic acid, dowell l 37, dequest 2000, nitrilotrimethylphosphonic acid, nitrilotris methylenephosphonic acid, aminotris methylphosphonic acid, aminotri methylenephosphonic acid | |
| [bis(phosphonomethyl)amino]methylphosphonic acid |
Specifications
| 6419-19-8 | |
| C3H12NO9P3 | |
| 25 mL | |
| YDONNITUKPKTIG-UHFFFAOYSA-N | |
| [bis(phosphonomethyl)amino]methylphosphonic acid | |
| 16698 | |
| Liquid |
| Yellow | |
| MFCD00002138 | |
| nitrilotris methylene triphosphonic acid, tris phosphonomethyl amine, ferrofos 509, aminotrimethylene phosphonic acid, dowell l 37, dequest 2000, nitrilotrimethylphosphonic acid, nitrilotris methylenephosphonic acid, aminotris methylphosphonic acid, aminotri methylenephosphonic acid | |
| C(N(CP(=O)(O)O)CP(=O)(O)O)P(=O)(O)O | |
| 299.048 | |
| 299.05 | |
| Nitrilotris(methylenephosphonic Acid) (ca. 50% in Water, ca. 2.2mol/L) |
Safety and Handling
EINECSNumber : (2)-1866&(2)-2009
RTECSNumber : SZ9860000
TSCA : Yes