missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrilotris(methylenephosphonic Acid) (ca. 50% in Water, ca. 2.2mol/L), TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 mL |
| Formula Weight | 299.05 |
| Physical Form | Liquid |
| Chemical Name or Material | Nitrilotris(methylenephosphonic Acid) (ca. 50% in Water, ca. 2.2mol/L) |
Safety and Handling
| EINECSNumber | (2)-1866&(2)-2009 |
| RTECSNumber | SZ9860000 |
| TSCA | Yes |