Learn More
Nitrilotriacetic acid trisodium salt monohydrate, 98+%
CAS: 18662-53-8 | C6H8NNa3O7 | 275.099 g/mol
$38.32 - $374.25
Chemical Identifiers
| CAS | 18662-53-8 |
|---|---|
| Molecular Formula | C6H8NNa3O7 |
| Molecular Weight (g/mol) | 275.099 |
| MDL Number | MFCD00150789 |
| InChI Key | HERBOKBJKVUALN-UHFFFAOYSA-K |
| Synonym | trisodium nitrilotriacetate monohydrate, nitrilotriacetic acid trisodium salt monohydrate, nta sodium hydrate, nta trisodium salt dihydrate, unii-j0u92u6nu5, ccris 437, nitrilotriacetic acid trisodium salt, monohydrate, nitriloacetic acid trisodium salt monohydrate, glycine, n,n-bis carboxymethyl-, trisodium salt, monohydrate, nitrilotriacetic acid, trisodium salt monohydrate |
| PubChem CID | 29194 |
| IUPAC Name | trisodium;2-[bis(carboxylatomethyl)amino]acetate;hydrate |
| SMILES | C(C(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.[Na+].[Na+].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1834822
|
Thermo Scientific Chemicals
A1834822 |
100 g |
Each for $38.32
|
|
|||||
|
AAA1834836
|
Thermo Scientific Chemicals
A1834836 |
500 g |
Each for $95.49
|
|
|||||
|
AAA183480E
|
Thermo Scientific Chemicals
A183480E |
2500 g |
Each for $374.25
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 18662-53-8 | |
| 275.099 | |
| HERBOKBJKVUALN-UHFFFAOYSA-K | |
| 29194 | |
| C(C(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.[Na+].[Na+].[Na+] |
| C6H8NNa3O7 | |
| MFCD00150789 | |
| trisodium nitrilotriacetate monohydrate, nitrilotriacetic acid trisodium salt monohydrate, nta sodium hydrate, nta trisodium salt dihydrate, unii-j0u92u6nu5, ccris 437, nitrilotriacetic acid trisodium salt, monohydrate, nitriloacetic acid trisodium salt monohydrate, glycine, n,n-bis carboxymethyl-, trisodium salt, monohydrate, nitrilotriacetic acid, trisodium salt monohydrate | |
| trisodium;2-[bis(carboxylatomethyl)amino]acetate;hydrate |
Specifications
| 18662-53-8 | |
| C6H8NNa3O7 | |
| 100 g | |
| 14,6579 | |
| HERBOKBJKVUALN-UHFFFAOYSA-K | |
| trisodium;2-[bis(carboxylatomethyl)amino]acetate;hydrate | |
| 29194 | |
| ≥98% |
| >300°C | |
| MFCD00150789 | |
| 3774022 | |
| trisodium nitrilotriacetate monohydrate, nitrilotriacetic acid trisodium salt monohydrate, nta sodium hydrate, nta trisodium salt dihydrate, unii-j0u92u6nu5, ccris 437, nitrilotriacetic acid trisodium salt, monohydrate, nitriloacetic acid trisodium salt monohydrate, glycine, n,n-bis carboxymethyl-, trisodium salt, monohydrate, nitrilotriacetic acid, trisodium salt monohydrate | |
| C(C(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.[Na+].[Na+].[Na+] | |
| 275.099 | |
| 275.12 (257.10 Anhydrous) | |
| Nitrilotriacetic acid trisodium salt monohydrate |
Safety and Handling
GHS H Statement
H301-H351-H319
Toxic if swallowed.
Suspected of causing cancer.
Causes serious eye irritation.
P201-P202-P264b-P270-P280-P301+P312-P305+P351+P338-P308+P313-P330-P501c
H302-H319-H351
EINECSNumber : 205-355-7
RTECSNumber : AJ1070000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only