missing translation for 'onlineSavingsMsg'
Learn More

Nitrilotriacetic acid trisodium salt monohydrate, 98+%

Catalog No. AAA1834836
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1834836 500 g
AAA1834822 100 g
AAA183480E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1834836 Supplier Thermo Scientific Chemicals Supplier No. A1834836

CAS: 18662-53-8 | C6H8NNa3O7 | 275.099 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18662-53-8
Molecular Formula C6H8NNa3O7
Molecular Weight (g/mol) 275.099
MDL Number MFCD00150789
InChI Key HERBOKBJKVUALN-UHFFFAOYSA-K
Synonym trisodium nitrilotriacetate monohydrate, nitrilotriacetic acid trisodium salt monohydrate, nta sodium hydrate, nta trisodium salt dihydrate, unii-j0u92u6nu5, ccris 437, nitrilotriacetic acid trisodium salt, monohydrate, nitriloacetic acid trisodium salt monohydrate, glycine, n,n-bis carboxymethyl-, trisodium salt, monohydrate, nitrilotriacetic acid, trisodium salt monohydrate
PubChem CID 29194
IUPAC Name trisodium;2-[bis(carboxylatomethyl)amino]acetate;hydrate
SMILES C(C(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.[Na+].[Na+].[Na+]

Specifications

Melting Point >300°C
Quantity 500 g
Beilstein 3774022
Merck Index 14,6579
Formula Weight 275.12 (257.10 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material Nitrilotriacetic acid trisodium salt monohydrate
Hazard Category H302-H319-H351
Hazard Statement GHS H Statement
H301-H351-H319
Toxic if swallowed.
Suspected of causing cancer.
Causes serious eye irritation.
Precautionary Statement P201-P202-P264b-P270-P280-P301+P312-P305+P351+P338-P308+P313-P330-P501c
EINECSNumber 205-355-7
RTECSNumber AJ1070000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only