missing translation for 'onlineSavingsMsg'
Learn More

Nitrilotriacetic Acid 98.0+%, TCI America™

Catalog No. N009825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N009825G 25 g
N0098500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N009825G Supplier TCI America Supplier No. N009825G
Only null left

Chemical Identifiers

CAS 139-13-9
Molecular Formula C6H9NO6
Molecular Weight (g/mol) 191.14
MDL Number MFCD00004287
InChI Key MGFYIUFZLHCRTH-UHFFFAOYSA-N
Synonym nitrilotriacetic acid, 2,2',2-nitrilotriacetic acid, triglycollamic acid, aminotriacetic acid, complexon i, trilon a, n,n-bis carboxymethyl glycine, nitrilotriacetate, komplexon i, titriplex i
PubChem CID 8758
ChEBI CHEBI:44557
IUPAC Name 2-[bis(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CC(O)=O)CC(O)=O

Specifications

Melting Point 245°C
Color White
pH 2.3 (saturated H2O soln.)
Quantity 25 g
UN Number 3077
Formula Weight 191.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nitrilotriacetic Acid
EINECSNumber (2)-1276
RTECSNumber AJ0175000
TSCA Yes