missing translation for 'onlineSavingsMsg'
Learn More

Nitrilotriacetic acid, Honeywell Fluka™

Catalog No. p-7072112
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 kg
50 g
Catalog No. Quantity
6002520 50 g
6002519 25 kg
2 options

Catalog No. 6002520

Supplier: Honeywell Chemicals 3453950G

for complexometry,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 139-13-9
Molecular Formula C6H9NO6
Molecular Weight (g/mol) 191.14
MDL Number MFCD00004287
InChI Key MGFYIUFZLHCRTH-UHFFFAOYSA-N
Synonym nitrilotriacetic acid, 2,2',2-nitrilotriacetic acid, triglycollamic acid, aminotriacetic acid, complexon i, trilon a, n,n-bis carboxymethyl glycine, nitrilotriacetate, komplexon i, titriplex i
PubChem CID 8758
ChEBI CHEBI:44557
IUPAC Name 2-[bis(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CC(O)=O)CC(O)=O

Specifications

Flash Point 100°C (212°F)
Linear Formula N(CH2COOH)3
Quantity 50 g
UN Number NONH for all modes of transport
Beilstein 1710776
Formula Weight 191.14g/mol
Percent Purity ≥99%
Chemical Name or Material Nitrilotriacetic acid
Hazard Statement P281-P305 + P351 + P338
RTECSNumber AJ0175000