missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrilotriacetic Acid, ACS Grade, ≥98%, MP Biomedicals™
$227.92 - $227.92
Chemical Identifiers
| CAS | 139-13-9 |
|---|---|
| Molecular Formula | C6H9NO6 |
| Molecular Weight (g/mol) | 191.14 |
| MDL Number | MFCD00004287 |
| InChI Key | MGFYIUFZLHCRTH-UHFFFAOYSA-N |
| Synonym | nitrilotriacetic acid, 2,2',2-nitrilotriacetic acid, triglycollamic acid, aminotriacetic acid, complexon i, trilon a, n,n-bis carboxymethyl glycine, nitrilotriacetate, komplexon i, titriplex i |
| PubChem CID | 8758 |
| ChEBI | CHEBI:44557 |
| IUPAC Name | 2-[bis(carboxymethyl)amino]acetic acid |
| SMILES | OC(=O)CN(CC(O)=O)CC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15255180
|
MP Biomedicals Inc
0215255180 |
100 g |
Each for $227.92
|
|
|||||
Chemical Identifiers
| 139-13-9 | |
| 191.14 | |
| MGFYIUFZLHCRTH-UHFFFAOYSA-N | |
| 8758 | |
| 2-[bis(carboxymethyl)amino]acetic acid |
| C6H9NO6 | |
| MFCD00004287 | |
| nitrilotriacetic acid, 2,2',2-nitrilotriacetic acid, triglycollamic acid, aminotriacetic acid, complexon i, trilon a, n,n-bis carboxymethyl glycine, nitrilotriacetate, komplexon i, titriplex i | |
| CHEBI:44557 | |
| OC(=O)CN(CC(O)=O)CC(O)=O |
Specifications
| 139-13-9 | |
| ≥98% | |
| MFCD00004287 | |
| nitrilotriacetic acid, 2,2',2-nitrilotriacetic acid, triglycollamic acid, aminotriacetic acid, complexon i, trilon a, n,n-bis carboxymethyl glycine, nitrilotriacetate, komplexon i, titriplex i | |
| OC(=O)CN(CC(O)=O)CC(O)=O | |
| 191.14 | |
| CHEBI:44557 | |
| ACS Reagent | |
| Nitrilotriacetic Acid |
| White | |
| C6H9NO6 | |
| 100 g | |
| MGFYIUFZLHCRTH-UHFFFAOYSA-N | |
| 2-[bis(carboxymethyl)amino]acetic acid | |
| 8758 | |
| 191.14 | |
| Solid |
Safety and Handling
Recommended Storage : Store at room temperature (15° to 30°C)