missing translation for 'onlineSavingsMsg'
Learn More

Nitrilotriacetic acid, 99%, Thermo Scientific™

Nitrilotriacetic acid, CAS # 139-13-9, is a tricarboxylic acid compound used for chemical chelation and elution in protein studies and the purification of certain earth compounds.

Supplier:  Thermo Scientific Chemicals 415740051

Catalog No. AC415740051

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General description

  • Nitrilotriacetic acid (2,2',2-nitrilotriacetic acid) is a tricarboxylic acid used for chemical purification


Applications

  • A known mutagen, suitable for cell culture experiments of DNA damage
  • Nitrilotriacetic acid can be used for protein elution using the His-tag method and chemical chelation
  • Nitrilotriacetic acid is capable of chelating Ca2+, Co2+, Cu2+, and Fe3+
  • Intended for sensitive applications such as HPLC
Specifications

Specifications

Nitrilotriacetic acid
139-13-9
100.0
99%
C6H9NO6
MFCD00004287
04,369
nitrilotriacetic acid, 2,2',2-nitrilotriacetic acid, triglycollamic acid, aminotriacetic acid, complexon i, trilon a, n,n-bis carboxymethyl glycine, nitrilotriacetate, komplexon i, titriplex i
MGFYIUFZLHCRTH-UHFFFAOYSA-N
2-[bis(carboxymethyl)amino]acetic acid
8758
191.14
99%
98.5
Authentic
Plastic drum
N(CH2CO2H)3
5kg
15,6665
Solubility in water: 1.28g/L (22.5°C). Other solubilities: soluble in most common organic solvents
OC(=O)CN(CC(O)=O)CC(O)=O
191.14
CHEBI:44557
99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Suspected of causing cancer if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy

GHS Signal Word: Warning

EINECSNumber : 205-355-7

RTECSNumber : AJ0175000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only