missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid/Dipicolinic Acid, IC eluent concentrate (10 x) for Metrosep C 6, MilliporeSigma™ Supelco™

Catalog No. p-7193372
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004773 1 L
111004463 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111004773 Supplier Merck Emd Millipore Supplier No. 193991L
Only null left

Concentrate HNO3 17 mM and Dipicolinic Acid 17 mM in H2O

This eluent concentrate is application tested. It is certified in accordance with ISO Guide 31. All details about exact content, uncertainty, traceability and expiry date are described in the certificate.

Specifications

Concentration 17 mM HNO 3 in. H2O
Quantity 1 L