missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nitrazine Yellow, indicator grade

Catalog No. AC190570050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC190570050 5 g
AC190570250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC190570050 Supplier Thermo Scientific Chemicals Supplier No. 190570050

CAS: 5424-01-1 | C5H5N3O2 | 139.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 1-1-5424
Molecular Formula C5H5N3O2
Molecular Weight (g/mol) 139.11
MDL Number MFCD00003938
InChI Key ZAGZIOYVEIDDJA-UHFFFAOYSA-N
Synonym 2-(2,4-Dinitrophenylazo)-1-naphthol-3,6-disulfonic acid, disodium salt, 3-(2, 4-Dinitrophenylazo)-4-hydroxy-2, 7-naphthalenedisulfonic acid, 2-(2, 4-Dinitrophenylazo)-1-naphthol-3, 6-disulfonic acid
PubChem CID 72656
IUPAC Name 3-aminopyrazine-2-carboxylic acid
SMILES C1=CN=C(C(=N1)C(=O)O)N

Specifications

Infrared Spectrum Authentic
Formula Weight 542.35
Grade Indicator
Packaging Glass bottle
UV_Vis 85% min.
Visual Transition Interval from pH 6.0(bright yellow) to pH 7.2(bright blue)
Color Brown to Orange
Quantity 5 g
Physical Form Powder
Chemical Name or Material Nitrazine yellow

RUO – Research Use Only