missing translation for 'onlineSavingsMsg'
Learn More

Nitrate Nitrogen Standard, For IC, Certified Reference Material, 1,000 mg/L N as Nitrate in H2O, MilliporeSigma™ Supelco™

Catalog No. 111005120
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111005120 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111005120 Supplier Merck Emd Millipore Supplier No. 53638100ML
Only null left

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.

Specifications

Product Line TraceCERT
Concentration 1000 mg/L N as nitrate aq. soln.
Quantity 100 mL
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC