missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Nitinol-core (NIT) SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB)

Catalog No. 111021711
Change view
Click to view available options
For Use With (Equipment):
Autosampler or Manual Holder
Manual Holder
Type:
23 ga
24 ga
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Type
111021711 Autosampler or Manual Holder 24 ga
111021708 Manual Holder 23 ga
111021709 Manual Holder 24 ga
111021710 Autosampler or Manual Holder 23 ga
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 111021711 Supplier MilliporeSigma™ Supelco™ Supplier No. 57931U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For analyte group volatiles, amines, and nitroaromatic compounds (MW 50-300)

  • Nitinol-core (NIT) SPME fibers were specifically engineered to be more reproducible and more robust than traditional SPME fibers that contain a fused silica (FS) core.
  • The NIT fibers offer > 30% improvement in lot to lot and fiber to fiber reproducibility when compared to the FS versions, but offer equivalent selectivity for easy transfer of methods from FS to the more robust, reproducible NIT version.

Specifications

For Use With (Equipment) Autosampler or Manual Holder
Type 24 ga
Film Thickness 65 μm
Material Blue (Metallic), Metal Alloy
Matrix Polydimethylsiloxane/Divinylbenzene (PDMS/DVB) coating
Quantity 3/Pk.