missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Nitinol-core (NIT) SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB)

For analyte group volatiles, amines, and nitroaromatic compounds (MW 50-300)

Supplier:  MilliporeSigma™ Supelco™ 57923U

 View more versions of this product

Catalog No. 111021710


Only null left

Description

Description

  • Nitinol-core (NIT) SPME fibers were specifically engineered to be more reproducible and more robust than traditional SPME fibers that contain a fused silica (FS) core.
  • The NIT fibers offer > 30% improvement in lot to lot and fiber to fiber reproducibility when compared to the FS versions, but offer equivalent selectivity for easy transfer of methods from FS to the more robust, reproducible NIT version.
Specifications

Chemical Identifiers

65 μm
Blue (Metallic), Metal Alloy
Polydimethylsiloxane/Divinylbenzene (PDMS/DVB) coating
3/Pk.

Specifications

Autosampler or Manual Holder
65 μm
Blue (Metallic), Metal Alloy
3/Pk.
23 ga
Solid phase microextraction (SPME): suitable
Polydimethylsiloxane/Divinylbenzene (PDMS/DVB) coating
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions