missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Nitinol-core (NIT) SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB)
For analyte group volatiles, amines, and nitroaromatic compounds (MW 50-300)
Supplier: MilliporeSigma™ Supelco™ 57921U
Description
- Nitinol-core (NIT) SPME fibers were specifically engineered to be more reproducible and more robust than traditional SPME fibers that contain a fused silica (FS) core.
- The NIT fibers offer > 30% improvement in lot to lot and fiber to fiber reproducibility when compared to the FS versions, but offer equivalent selectivity for easy transfer of methods from FS to the more robust, reproducible NIT version.
Chemical Identifiers
| 65 μm | |
| Blue (Metallic), Metal Alloy | |
| Polydimethylsiloxane/Divinylbenzene (PDMS/DVB) coating | |
| 3/Pk. |
Specifications
| Manual Holder | |
| 65 μm | |
| Blue (Metallic), Metal Alloy | |
| 3/Pk. |
| 24 ga | |
| Solid phase microextraction (SPME): suitable | |
| Polydimethylsiloxane/Divinylbenzene (PDMS/DVB) coating |