missing translation for 'onlineSavingsMsg'
Learn More

Nistose Trihydrate 99.0+%, TCI America™

Catalog No. N05711G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N05711G Supplier TCI America Supplier No. N05711G
Only null left

Chemical Identifiers

CAS 13133-07-8
Molecular Formula C24H42O21
Molecular Weight (g/mol) 666.579
MDL Number MFCD00191672
InChI Key FLDFNEBHEXLZRX-DLQNOBSRSA-N
Synonym Fungitetraose, Nystose
PubChem CID 166775
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-2-[[(2R,3S,4S,5R)-2-[[(2R,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxymethyl]-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]oxymethyl]-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,
SMILES C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)COC3(C(C(C(O3)CO)O)O)COC4(C(C(C(O4)CO)O)O)CO)O)O)O)O

Specifications

Color White
Quantity 1 g
Formula Weight 666.58
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Nistose Trihydrate