missing translation for 'onlineSavingsMsg'
Learn More

NOTRAX™ NIRU™ Cushion-Ease™ GSII™ Matting System

Niru™ Cushion-Ease™ GSII™ is a high performance, modular, anti-fatigue matting system with a drainage top anti-slip surface texture and a multi-nib backing for comfort and ergonomic support

$277.24 - $462.08

Specifications

Description Multi-nib backing
Color Black
Length (English) 3 ft.
Quantity 1 Ea.
Thickness (English) 3/4 in.
View More Specs

Products 2
Catalog Number Mfr. No. Length (English) Width (English) Price Quantity  
Catalog Number Mfr. No. Length (English) Width (English) Price Quantity  
19086190
View Documents
NOTRAX™
850S0033BL
3 ft. 3 ft.
Each for $277.24
Only null left
 
19086191
View Documents
NOTRAX™
850S0035BL
3 ft. 5 ft.
Each for $462.08
Only null left
 
Description

Description

  • Resilient rubber compound and unique multi-nib support design offers exceptional fatigue relief
  • Made of nitrile rubber compound that provides resistance to chemicals, cutting fluids, animal fats, oils and greases
  • Large hole drainage design keeps moisture and debris free away
  • Easy to snap tog system for on-site custom work station platforms
  • Nitrile provides highest level of resistance to chemicals, oils and greases
  • GSII™ offers the ultimate in durability, performance and traction in harsh environments
  • Compatible with any Cushion-Ease™ family mats
  • 551 M.D. Ramp System™ and 552 extra wide M.D.-X Ramp System™ available
  • 850FR fire retardant is available
Specifications

Specifications

Multi-nib backing
3 ft.
3/4 in.
Black
1 Ea.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'