Learn More
Nipecotic acid, 98%
CAS: 498-95-3 | C6H11NO2 | 129.16 g/mol
Supplier: Thermo Scientific Chemicals B2472306
Description
GABA reuptake inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Nipecotic acid | |
| ∼261°C (decomposition) | |
| MFCD00005992 MFCD01630787 | |
| 81096 | |
| nipecotic acid, 3-piperidinecarboxylic acid, hexahydronicotinic acid, +/--nipecotic acid, h-dl-nip-oh, +/--3-piperidine carboxylic acid, dl-piperidine-3-carboxylic acid, nipecotic, h-nip-oh, 3-carboxypiperidine | |
| XJLSEXAGTJCILF-UHFFFAOYSA-N | |
| piperidine-3-carboxylic acid | |
| 4498 | |
| 129.16 |
| 498-95-3 | |
| C6H11NO2 | |
| 5 g | |
| 14,6561 | |
| Soluble in water | |
| OC(=O)C1CCCNC1 | |
| 129.16 | |
| CHEBI:116931 | |
| 98% |
Chemical Identifiers
| 498-95-3 | |
| 129.16 | |
| XJLSEXAGTJCILF-UHFFFAOYSA-N | |
| 4498 | |
| piperidine-3-carboxylic acid |
| C6H11NO2 | |
| MFCD00005992 MFCD01630787 | |
| nipecotic acid, 3-piperidinecarboxylic acid, hexahydronicotinic acid, +/--nipecotic acid, h-dl-nip-oh, +/--3-piperidine carboxylic acid, dl-piperidine-3-carboxylic acid, nipecotic, h-nip-oh, 3-carboxypiperidine | |
| CHEBI:116931 | |
| OC(=O)C1CCCNC1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 207-873-9
RTECSNumber : TM6125380
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only