missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Niobium wire, 0.25mm (0.01in) dia, 99.99% (metals basis excluding Ta)
CAS: 7440-03-1 | Nb | 92.91 g/mol
Supplier: Thermo Scientific Chemicals 010265BY
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Niobium wire | |
| 3-1-7440 | |
| Odorless | |
| Wire | |
| Nb | |
| columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio | |
| [Nb] | |
| 92.91 | |
| CHEBI:33344 |
| 0.25mm (0.01 in.) dia. | |
| 100 cm | |
| ≈0.42g/m | |
| (metals basis excluding Ta) | |
| MFCD00011126 | |
| GUCVJGMIXFAOAE-UHFFFAOYSA-N | |
| niobium | |
| 23936 | |
| 99.99% |
Chemical Identifiers
| 3-1-7440 | |
| 92.91 | |
| GUCVJGMIXFAOAE-UHFFFAOYSA-N | |
| 23936 | |
| niobium |
| Nb | |
| MFCD00011126 | |
| columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio | |
| CHEBI:33344 | |
| [Nb] |
Safety and Handling
EINECSNumber : 231-113-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only