Learn More
Niobium(V) oxide, 99.99%, (trace metal basis)
CAS: 1313-96-8 | Nb2O5 | 265.81 g/mol
$78.34 - $239.66
Chemical Identifiers
| CAS | 1313-96-8 |
|---|---|
| Molecular Formula | Nb2O5 |
| Molecular Weight (g/mol) | 265.81 |
| MDL Number | MFCD00011128 |
| InChI Key | ZKATWMILCYLAPD-UHFFFAOYSA-N |
| Synonym | niobium oxide, niobium v oxide, niobium pentoxide, diniobium pentaoxide, niobia, niobium pentaoxide, diniobium pentoxide, niobium 5+ oxide, nobum oxde |
| PubChem CID | 123105 |
| SMILES | O=[Nb](=O)O[Nb](=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC193630100
|
Thermo Scientific Chemicals
193630100 |
10 g | Glass bottle |
Each for $78.34
|
|
||||
|
AC193630500
|
Thermo Scientific Chemicals
193630500 |
50 g | Plastic bottle |
Each for $239.66
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisChemical Identifiers
| 1313-96-8 | |
| 265.81 | |
| ZKATWMILCYLAPD-UHFFFAOYSA-N | |
| 123105 |
| Nb2O5 | |
| MFCD00011128 | |
| niobium oxide, niobium v oxide, niobium pentoxide, diniobium pentaoxide, niobia, niobium pentaoxide, diniobium pentoxide, niobium 5+ oxide, nobum oxde | |
| O=[Nb](=O)O[Nb](=O)=O |
Specifications
| 1520.0°C | |
| White | |
| 100.0 | |
| 10 g | |
| Nb2O5 | |
| MFCD00011128 | |
| niobium oxide, niobium v oxide, niobium pentoxide, diniobium pentaoxide, niobia, niobium pentaoxide, diniobium pentoxide, niobium 5+ oxide, nobum oxde | |
| ZKATWMILCYLAPD-UHFFFAOYSA-N | |
| dioxoniobiooxy(dioxo)niobium | |
| 123105 | |
| 99.99% | |
| Glass bottle | |
| 4.4700g/mL |
| 1313-96-8 | |
| 99.95 | |
| Powder | |
| 99.99% | |
| Nb2O5 | |
| 15, 6645 | |
| Solubility in water: insoluble. Other solubilities: soluble in hf and hot h2so4, soluble in fused potassium hydrogen sulfate or, carbonates or hydroxides of the alkali metals | |
| O=[Nb](=O)O[Nb](=O)=O | |
| 265.81 | |
| 265.8 | |
| Trace Metal Basis | |
| 4.47 | |
| Niobium(V) oxide |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 215-213-6
RUO – Research Use Only